EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Synthesis of 4β-carbamoyl epipodophyllotoxins as potential antitumour agents.
PMID 21489802
Abstract
A series of new 4β-carbamoyl epipodophyllotoxin analogues have been synthesized and evaluated for their anticancer activity against eleven cancer cell lines including Zr-75-1, MCF7, KB, Gurav, DWD, Colo 205, A-549, Hop62, PC3, SiHa and A-2780. Most of the compounds exhibited better growth-inhibition activities against tested cell lines than that of etoposide. Further, compounds 6g and 6i are also evaluated for their DNA topoisomerase-II (topo-II) inhibition activity and they exhibited significant inhibition of topo-II catalytic activity comparable to etoposide.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
246557
Tryptamine hydrochloride, 99%
C10H12N2 · HCl