EMAIL THIS PAGE TO A FRIEND
Muscle & nerve
Topical naphazoline in the treatment of myasthenic blepharoptosis.
PMID 21491460
Abstract
When treating ocular myasthenia gravis (MG), the risk/benefit profile of corticosteroids is unclear, and acetylcholinesterase inhibitors are not very effective. We examined the efficacy of topical naphazoline in the treatment of myasthenic blepharoptosis. Sixty MG patients with blepharoptosis (32 with ocular symptoms only and 28 with mild generalized symptoms) were enrolled in a multicenter open trial of topical naphazoline. The effects were reported by patients via a questionnaire and were also confirmed for each patient at the clinic. Among 70 eyes of 60 patients, 20 eyes (28.6%) of 17 patients (28.3%) exhibited a marked response (full eye opening), and 24 eyes (34.3%) of 20 patients (33.3%) showed a good response (adequate but incomplete eye opening). Topical naphazoline was evaluated as useful in the treatment of myasthenic blepharoptosis by >70% of the patients. Topical naphazoline was found to be an effective supplementary symptomatic treatment for myasthenic blepharoptosis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N5504
Naphazoline hydrochloride
C14H14N2 · HCl