EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Screening for compounds with aromatase inhibiting activities from Atractylodes macrocephala Koidz.
PMID 21494203
Abstract
Ten compounds were isolated from the dichloromethane extract of Atractylodes macrocephala and their aromatase inhibiting activities were tested using an in vitro fluorescent-based aromatase assay. The results indicated that atractylenolide I, atractylenolide II and atractylenolide III had inhibition ratios of 94.56 ± 0.70%, 90.93 ± 1.41% and 86.31 ± 8.46%, respectively, at a concentration of 10 μM. We conclude from our results that atractylenolide and its derivates may serve as potential aromatase inhibitors (AIs) and thus merit continued study in the future.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A2737 Atractylenolide I, ≥98% (HPLC)
C15H18O2
A2987
Atractylenolide III, ≥98% (HPLC)
C15H20O3