EMAIL THIS PAGE TO A FRIEND
Revista medico-chirurgicala a Societatii de Medici si Naturalisti din Iasi
[Separation of erythromycin by direct pertraction from simulated broths].
PMID 21495378
Abstract
Erythromycin has been separated from simulated broths by free and facilitated pertraction with D2EHPA dissolved in dichloromethane using an U-shaped pertraction cell. The pertraction has been analyzed by means of initial and final mass flows and permeability factor. The main factors which control the pertraction efficiency were identified to be the pH-gradient between the feed and stripping phase, aqueous phases viscosities and mixing intensity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
237825
Bis(2-ethylhexyl) phosphate, 97%
C16H35O4P