EMAIL THIS PAGE TO A FRIEND
Equine Veterinary Journal
Multiple immunofluorescence labelling enables simultaneous identification of all mature fibre types in a single equine skeletal muscle cryosection.
PMID 21496090