EMAIL THIS PAGE TO A FRIEND
Traffic (Copenhagen, Denmark)
Regulation of β-catenin nuclear dynamics by GSK-3β involves a LEF-1 positive feedback loop.
PMID 21496192
Abstract
Nuclear localization of β-catenin is integral to its role in Wnt signaling and cancer. Cellular stimulation by Wnt or lithium chloride (LiCl) inactivates glycogen synthase kinase-3β (GSK-3β), causing nuclear accumulation of β-catenin and transactivation of genes that transform cells. β-catenin is a shuttling protein; however, the mechanism by which GSK-3β regulates β-catenin nuclear dynamics is poorly understood. Here, fluorescence recovery after photobleaching assays were used to measure the β-catenin-green fluorescent protein dynamics in NIH 3T3 cells before and after GSK-3β inhibition. We show for the first time that LiCl and Wnt3a cause a specific increase in β-catenin nuclear retention in live cells and in fixed cells after detergent extraction. Moreover, LiCl reduced the rate of nuclear export but did not affect import, hence biasing β-catenin transport toward the nucleus. Interestingly, the S45A mutation, which blocks β-catenin phosphorylation by GSK-3β, did not alter nuclear retention or transport, implying that GSK-3β acts through an independent regulator. We compared five nuclear binding partners and identified LEF-1 as the key mediator of Wnt3a and LiCl-induced nuclear retention of β-catenin. Thus, Wnt stimulation triggered a LEF-1 positive feedback loop to enhance the nuclear chromatin-retained pool of β-catenin by 100-300%. These findings shed new light on regulation of β-catenin nuclear dynamics.
Related Materials