EMAIL THIS PAGE TO A FRIEND
Clinics in dermatology
Current dilemmas and controversies in allergic contact dermatitis to ophthalmic medications.
PMID 21496737
Abstract
Identifying contact allergens in ophthalmic medications can be a challenging and daunting experience. We summarize data on topical ophthalmic medications with the potential to cause periorbital contact dermatitis and allergic conjunctivitis, highlighting current dilemmas and controversies in this area. The following groups of allergens are reviewed: preservatives, antiglaucoma medications (prostaglandin analogues, β-blockers, carbonic anhydrase inhibitors, parasympathomimetics, sympathomimetics), antiinflammatory medications (nonsteroidal antiinflammatory drugs, corticosteroids), antibiotics, antivirals, antiallergic medications (antihistamines, cromones), anaesthetics, mydriatics, and cycloplegics.