EMAIL THIS PAGE TO A FRIEND
Veterinary microbiology
Determination of minimum inhibitory and minimum bactericidal concentrations of tiamulin against field isolates of Actinobacillus pleuropneumoniae.
PMID 21497460
Abstract
Tiamulin activity was measured against 19 UK field isolates of Actinobacillus pleuropneumoniae collected between 2003 and 2009 and the type strain ATCC 27090 as a control, with the intention of comparing broth with serum as growth media. Broth microdilution MIC/MBC tests were performed in accordance with the Clinical and Laboratory Standards Institute (CLSI) guideline M31-A3, in 'Veterinary Fastidious Medium' (VFM) (supplemented Mueller-Hinton broth at pH 7.3) and in 100% swine serum. For improved precision, a modified, overlapping doubling-dilution series was used (tiamulin concentration range 0.3-72 μg/ml). The MBC was reported as the lowest concentration producing a 99.9% reduction in bacterial density in the sub-cultured well contents, relative to the starting inoculum. The mean MBC/MIC ratio for tiamulin against A. pleuropneumoniae in VFM was low (1.74:1), even though tiamulin is classed as a bacteriostatic drug. Only three of the 19 isolates and the reference strain grew in 100% serum and their MICs were higher than those determined in VFM. It is postulated that this difference was due to differences in pH of the matrices or binding of tiamulin to serum proteins or a combination of both factors.