EMAIL THIS PAGE TO A FRIEND
Graefe's archive for clinical and experimental ophthalmology = Albrecht von Graefes Archiv fur klinische und experimentelle Ophthalmologie
Timolol versus brinzolamide added to travoprost in glaucoma or ocular hypertension.
PMID 21499770
Abstract
To compare the efficacy and safety of timolol 0.5% versus brinzolamide 1.0% when added to travoprost monotherapy in patients with primary open-angle glaucoma or ocular hypertension. Patients meeting selection criteria (IOP one eye 19 mmHg and ≤32 mmHg and IOP both eyes ≤32 mmHg at 8:00 h) were switched to travoprost monotherapy for 4 weeks. Patients then insufficiently controlled on travoprost (IOP at 8:00 h ≥19 mmHg) at baseline were randomized to receive either travoprost and brinzolamide or travoprost and timolol in a double-masked fashion for 12 weeks. Two hundred and fifty-three patients underwent the 4-week run-in period. Switching to travoprost resulted in adequate IOP control (<19 mmHg) for 21.7% of patients. After 3 months of treatment, both drug combinations statistically significantly reduced the mean IOP at each time point (8:00, 12:00 and 16:00 h) and the mean diurnal IOP, which was 17.9 ± 2.6 mmHg for the brinzolamide group and 17.0 ± 3.2 mmHg for the timolol group. Both combinations were well-tolerated. However, a statistically significant difference occurred at 16:00 h, with pressures of 16.4 ± 3.2 mmHg and 17.3 ± 2.8 mmHg for the timolol and brinzolamide groups, respectively (p = 0.038). Fifty percent of patients reported one adverse event, whereas in 13.2% three or more adverse effects were named. Hyperemia was found most often (6.3% of the patients). Both adjunctive combinations moderately reduced IOP in patients inadequately controlled with travoprost monotherapy, with timolol being slightly stronger 8 hours after instillation. Adjunctive treatment with brinzolamide and travoprost may be an alternative for patients not tolerant or not responsive to treatment with timolol and travoprost.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0216
Brinzolamide, ≥98% (HPLC)
C12H21N3O5S3