EMAIL THIS PAGE TO A FRIEND
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes
Concurrent 2,4-D and triclopyr biomonitoring of backpack applicators, mixer/loader and field supervisor in forestry.
PMID 21500074
Abstract
Two herbicides, 2,4-D and triclopyr esters (application ratio 1.6:1 acid equivalents) were applied as a tank mix by a crew of 8 backpack sprayer applicators, a mixer/loader, and a field supervisor. The crew was employed in a conifer release program in northern California during the summer of 2002. Biomonitoring (urine, 24 h) utilized 2,4-D and triclopyr (a.e.) as rapidly excreted exposure biomarkers. The absorbed dosages of 2,4-D and triclopyr were calculated based upon cotton whole body suits and biomonitoring. Dosages based upon accumulation of the herbicides on body suits averaged 42.6 μg (a.e.) 2,4-D/kg-d and 8.0 μg (a.e.) triclopyr/kg-d. Six consecutive days of concurrent urine collections showed that backpack applicators excreted an average of 11.0 μg (a.e.) 2,4-D/kg-d and 18.9 μg (a.e.) triclopyr/kg-d. Estimates based upon curve fitting were 17.1 and 29.3 μg (a.e.)/kg-d, respectively. Results suggest that passive dosimetry for 2,4-D consistently overestimated the dosage measured using biomonitoring by a factor of 2-3 fold, while for triclopyr, passive dosimetry underestimated the absorbed dose based on biomonitoring by a factor of 2-4 fold.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32016
Triclopyr, PESTANAL®, analytical standard
C7H4Cl3NO3