EMAIL THIS PAGE TO A FRIEND
Topics in current chemistry
Rhodium-catalyzed C-H bond arylation of arenes.
PMID 21500409
Abstract
A review is presented of synthetic methods for the preparation of biaryls by the rhodium-catalyzed C-H bond arylation of arenes with aryl halides (C-H/ C-X couplings), arylmetal reagents (C-H/C-M couplings) and arenes (C-H/C-H couplings), with an emphasis on postulated mechanisms and their implications on reactivity, selectivity and substrate scope.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
357340
Rhodium, foil, thickness 0.025 mm, 99.9% trace metals basis
Rh
204218
Rhodium, powder, 99.95% trace metals basis
Rh
267368
Rhodium, sponge, 99.9% trace metals basis
Rh
GF94930667
Rhodium, microfoil, disks, 10mm, thinness 0.25μm, specific density 310μg/cm2, permanent mylar 3.5μm support, 99.99%
Rh
GF64213680
Rhodium, wire reel, 0.05m, diameter 0.12mm, as drawn, 99.9%
Rh
GF37380089
Rhodium, wire reel, 0.2m, diameter 0.25mm, hard, 99.9%
Rh
GF45819995
Rhodium, wire reel, 0.05m, diameter 0.25mm, hard, 99.9%
Rh
GF83497652
Rhodium, wire reel, 0.05m, diameter 1.0mm, as drawn, 99.9%
Rh
GF26710680
Rhodium, wire reel, 0.1m, diameter 0.12mm, as drawn, 99.9%
Rh
GF27499045
Rhodium, wire reel, 0.1m, diameter 0.25mm, hard, 99.9%
Rh
GF06539979
Rhodium, wire reel, 0.1m, diameter 0.5mm, as drawn, 99.9%
Rh
GF37207166
Rhodium, wire reel, 0.1m, diameter 1.0mm, as drawn, 99.9%
Rh
GF38835737
Rhodium, wire reel, 0.2m, diameter 0.12mm, as drawn, 99.9%
Rh
GF17496973
Rhodium, wire reel, 0.2m, diameter 0.5mm, as drawn, 99.9%
Rh
GF54466937
Rhodium, wire reel, 0.5m, diameter 0.12mm, as drawn, 99.9%
Rh
GF72510364
Rhodium, wire reel, 0.5m, diameter 0.25mm, hard, 99.9%
Rh
GF44569907
Rhodium, foil, 25x25mm, thickness 0.125mm, as rolled, 99.9%
Rh
GF80841146
Rhodium, foil, 25x25mm, thickness 0.15mm, as rolled, 99.9%
Rh
GF40539405
Rhodium, foil, 25x25mm, thickness 0.1mm, as rolled, 99.9%
Rh
GF20569697
Rhodium, foil, 12mm disks, thickness 0.5mm, as rolled, 99.9%
Rh
GF36379345
Rhodium, foil, light tested, 15x15mm, thickness 0.05mm, as rolled, 99.9%
Rh
GF57675561
Rhodium, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99.9%
Rh
GF12568013
Rhodium, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, x-ray quality, 99.9%
Rh
GF96356559
Rhodium, foil, light tested, 25x25mm, thickness 0.075mm, as rolled, 99.9%
Rh
GF04978963
Rhodium, foil, not light tested, 10x10mm, thickness 0.012mm, as rolled, 99.9%
Rh
GF17514312
Rhodium, foil, not light tested, 25x25mm, thickness 0.00025mm, permanent polyester support, 99.9%
Rh
GF35442727
Rhodium, foil, not light tested, 25x25mm, thickness 0.0005mm, permanent polyester support, 99.85%
Rh
GF49036204
Rhodium, foil, not light tested, 25x25mm, thickness 0.00075mm, permanent polyester support, 99.85%
Rh
GF87742659
Rhodium, foil, not light tested, 25x25mm, thickness 0.001mm, permanent polyester support, 99.85%
Rh
GF15499413
Rhodium, foil, not light tested, 25x25mm, thickness 0.006mm, as rolled, 99.9%
Rh