EMAIL THIS PAGE TO A FRIEND
Journal of endocrinological investigation
Early hypertension and prolonged mineralocorticoid therapy discontinuation in a child with salt-wasting 21-hydroxylase deficiency.
PMID 21502797
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F6127
Fludrocortisone acetate, ≥98%
C23H31FO6