EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Novel anti-Prelog stereospecific carbonyl reductases from Candida parapsilosis for asymmetric reduction of prochiral ketones.
PMID 21505708
Abstract
The application of biocatalysis to the synthesis of chiral molecules is one of the greenest technologies for the replacement of chemical routes due to its environmentally benign reaction conditions and unparalleled chemo-, regio- and stereoselectivities. We have been interested in searching for carbonyl reductase enzymes and assessing their substrate specificity and stereoselectivity. We now report a gene cluster identified in Candida parapsilosis that consists of four open reading frames including three putative stereospecific carbonyl reductases (scr1, scr2, and scr3) and an alcohol dehydrogenase (cpadh). These newly identified three stereospecific carbonyl reductases (SCRs) showed high catalytic activities for producing (S)-1-phenyl-1,2-ethanediol from 2-hydroxyacetophenone with NADPH as the coenzyme. Together with CPADH, all four enzymes from this cluster are carbonyl reductases with novel anti-Prelog stereoselectivity. SCR1 and SCR3 exhibited distinct specificities to acetophenone derivatives and chloro-substituted 2-hydroxyacetophenones, and especially very high activities towards ethyl 4-chloro-3-oxobutyrate, a β-ketoester with important pharmaceutical potential. Our study also showed that genomic mining is a powerful tool for the discovery of new enzymes.