EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Palladium-catalyzed double C-H activation directed by sulfoxides in the synthesis of dibenzothiophenes.
PMID 21506222
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
347833
Dibenzothiophene, ≥99%
C12H8S
45776
Dibenzothiophene, analytical standard
C12H8S