EMAIL THIS PAGE TO A FRIEND
Fertility and sterility
Association of sperm morphology and the sperm deformity index (SDI) with poly (ADP-ribose) polymerase (PARP) cleavage inhibition.
PMID 21507396
Abstract
Apoptosis was induced in immature and mature sperm in the presence or absence of poly (ADP-ribose) polymerase (PARP) inhibitor. The association of cleaved (cPARP) with sperm morphology was examined using sperm deformity index (SDI) score. The SDI scores are associated with PARP cleavage as an early marker of apoptosis.