EMAIL THIS PAGE TO A FRIEND
Psychopharmacology
Inhibition of phosphodiesterase-4 decreases ethanol intake in mice.
PMID 21509503
Abstract
Cyclic AMP (cAMP)-protein kinase A signaling has been implicated in the regulation of ethanol consumption. Phosphodiesterase-4 (PDE4) specifically hydrolyzes cAMP and plays a critical role in controlling intracellular cAMP levels in the brain. However, the role of PDE4 in ethanol consumption remains unknown. The objective of this study is to examine whether PDE4 was involved in regulating ethanol intake. The two-bottle choice paradigm was used to assess intake of ethanol, sucrose, and quinine in C57BL/6J mice treated with the selective PDE4 inhibitor rolipram or Ro 20-1724; locomotor activity was also monitored using the open-field test in mice treated with rolipram. Administration (i.p.) of either rolipram (0.25 and 0.5 mg/kg) or Ro 20-1724 (10 mg/kg) reduced ethanol intake and preference by 60-80%, but did not alter total fluid intake. In contrast, rolipram even at the higher dose of 0.5 mg/kg was not able to affect intake of sucrose or quinine, alcohol-induced sedation, or blood ethanol elimination. At 0.5 mg/kg, rolipram did decrease locomotor activity, but the effect only lasted for approximately 40 min, which did not likely affect behavior of ethanol drinking. These results suggest that PDE4 is a novel target for drugs that reduce ethanol intake; PDE4 inhibitors may be used for treatment of alcohol dependence.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B8279
4-(3-Butoxy-4-methoxybenzyl)imidazolidin-2-one, solid
C15H22N2O3