EMAIL THIS PAGE TO A FRIEND
Bioresource technology
Towards industrially-feasible delignification and pitch removal by treating paper pulp with Myceliophthora thermophila laccase and a phenolic mediator.
PMID 21511459
Abstract
The ability of two natural phenols to act as mediators of the recombinant Myceliophthora thermophila laccase (MtL) in eucalypt-pulp delignification was investigated. After alkaline peroxide extraction, the properties of the enzymatically-treated pulps improved with respect to the control. The pulp brightness increased (3.1 points) after the enzymatic treatment with MtL alone, but the highest improvements were obtained after the MtL treatment using syringaldehyde (4.7 points) and especially methyl syringate (8.3 points) as mediators. Likewise, a decrease in kappa number up to 2.7 points was obtained after the MtL-methyl syringate treatment, followed by decreases of 1.4 and 0.9 points after the treatments with MtL-syringaldehyde and MtL alone, respectively. On the other hand, removal of the main lipophilic extractives present in eucalypt pulp was observed after the above laccase-mediator treatments. Finally, the doses of both MtL and methyl syringate were reduced, and results compatible with industrial implementation were obtained.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S7602
Syringaldehyde, 98%
C9H10O4
W404926
Syringaldehyde, ≥98%, FG
C9H10O4