EMAIL THIS PAGE TO A FRIEND
Bioresource technology
Purification, amino acid sequence and characterization of the class IIa bacteriocin weissellin A, produced by Weissella paramesenteroides DX.
PMID 21511463
Abstract
Weissella paramesenteroides DX has been shown to produce a 4450-Da class IIa bacteriocin, weissellin A, composed of 43 amino acids with the sequence KNYGNGVYCNKHKCSVDWATFSANIANNSVAMAGLTGGNAGN. The bacteriocin shares 68% similarity with leucocin C from Leuconostoc mesenteroides. Computational analyses predict that the bacteriocin is a hydrophobic molecule with a beta-sheet type conformation. Weissellin A exhibited various levels of activity against all gram-positive bacteria tested, but was not active against Salmonella enterica Enteritidis. The antimicrobial activity was not associated with target-cell lysis. The bacteriocin retained activity after exposure to 121°C for 60 min or to -20°C for 6months, and to pH 2.0-10.0. It was not sensitive to trypsin, α-chymotrypsin, pepsin and papain, but was inactivated by proteinase K. At a dissolved oxygen concentration of 50%, weissellin A was produced with growth-associated kinetics. The properties of weissellin A make this bacteriocin a potentially suitable agent for food and feed preservation.