EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Synthesis of oxabicyclo[3.3.1]nonenes and substituted tetrahydropyrans via (3,5)-oxonium-ene reaction.
PMID 21512689
Abstract
An efficient methodology for the synthesis of oxabicyclo[3.3.1]nonenes and substituted tetrahydropyrans has been developed in moderate yields from the reaction of geraniol with aldehydes and epoxides promoted by boron trifluoride etherate.