EMAIL THIS PAGE TO A FRIEND
European journal of anaesthesiology
Providing effective implementation of antiemetic strategies: the postoperative nausea and vomiting-free hospital is a laudable and realistic goal.
PMID 21513079
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P6402
Perphenazine
C21H26ClN3OS