EMAIL THIS PAGE TO A FRIEND
Leukemia research
PTPIP51 is phosphorylated by Lyn and c-Src kinases lacking dephosphorylation by PTP1B in acute myeloid leukemia.
PMID 21513978
Abstract
Protein tyrosine phosphatase interacting protein 51 (PTPIP51) is known to be expressed in blood cells with restriction to the myeloid lineage. All myeloid progenitor cells are PTPIP51 positive except for the myeloblasts. To define the expression of PTPIP51 in acute myeloid leukemia (AML), we performed immunohistochemical experiments with peptide specific antibodies (C-terminus, N-terminus and aas 114-129) to PTPIP51 with samples of AML bone marrow trephine biopsy specimens. AML blasts reacted positive for PTPIP51 protein encompassing the C-terminal sequence. Healthy bone marrow displayed an exclusive staining for the N-terminal containing form of PTPIP51. Moreover, PTPIP51 protein was highly phosphorylated at its tyrosine 176 residue. Acquired confocal images of AML cells displayed an absence of PTP1B and revealed a co-localization of PTPIP51 and Lyn. Duolink proximity ligation assays (DPLA) corroborated an interaction for PTPIP51 with Lyn and c-Src. In AML blasts rarely an interaction of PTPIP51 with PTP1B and Raf-1 was seen. Furthermore, DPLA signals were also obtained for PTPIP51 and c-Kit in AML cells. Therefore, PTPIP51 was identified as a new signal molecule of the c-Kit signaling pathway. By the phosphorylation done by Lyn, c-Src and c-Kit, PTPIP51 is prevented to influence mitogen activated protein kinase pathway on Raf-1 level contributing to increased proliferation of AML cells.