EMAIL THIS PAGE TO A FRIEND
Journal of hazardous materials
Phosphorus removal performance of acid mine drainage from wastewater.
PMID 21514994
Abstract
Acid mine drainage (AMD) in Yunfu iron sulfide mine contain Fe(2+), Fe(3+), and Al(3+) up to 8000, 1700 and 1200 mg/L, respectively. Phosphorus removal from synthetic wastewater with 10mg/L of total phosphorus (TP) concentration and second municipal effluent with 3.5-4.0mg/L of TP concentration were conducted with the AMD by jar tests. Dosage of the AMD and initial pH of water are the two most important parameters affecting the performance of phosphorus removal of the AMD. The optimal phosphorus removal efficiency and residual iron ions (TFe) concentration are 97.0% and 3.0mg/L, respectively, at 1.61 Fe/P molar ratio and pH 8.03 for synthetic wastewater, and 92.1% and 0.32 mg/L, respectively, for second municipal effluent at 1.41 Fe/P molar ratio and pH 7.3. Resultant heavy metal concentration in effluents and precipitate was very low, and the risk of resultant heavy metal contamination was very small. The phosphorus removal performance of the AMD was much similar to that of ferric sulfate (FS) and polyferric sulfate (PFS), and better than that of FeSO(4). And residual TFe concentration in treated water arising from utilization of the AMD was similar to that of FeSO(4), and higher than that of FS and PFS. The AMD could be used as coagulant for phosphorus removal from wastewater directly due to the presence of Fe(2+), Fe(3+), and Al(3+) largely.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F0638
Iron(III) sulfate hydrate, suitable for plant cell culture, BioReagent
Fe2O12S3 · xH2O
307718
Iron(III) sulfate hydrate, 97%
Fe2O12S3 · xH2O