EMAIL THIS PAGE TO A FRIEND
Journal of hazardous materials
Evaluation of acute toxicity and teratogenic effects of plant growth regulators by Daphnia magna embryo assay.
PMID 21514995
Abstract
This study selected common plant growth regulators (Atonik, Cytokinin, Ethephon, Gibberellic acid and Paclobutrazol) to investigate their biological toxicity to the waters of the important biological indicator Daphnia magna. The methods used in this study included traditional neonate acute toxicity test, new Daphnia embryo toxicity test, and teratogenic embryo test. The study concluded that the acute toxicity of the five PGRs to Daphnia neonate had EC(50) value range of 1.9-130.5 mg l(-1), while acute toxicity of PGRs on Daphnia embryo had EC(50) value range of 0.2-125 mg l(-1); the Daphnia embryos' LOEC values (0.05-48 mg l(-1)) for the five PGRs were lower than embryo EC(50) values. The toxic ratios of 48 h EC(50) (neonate)/48 h LOEC (embryo) for 5 PGRs were 19-512 times. The study found that teratogenic effects of Paclobutrazol and Cytokinin induced in embryo were higher than those of most other PGRs. Microscopic observation of the teratogenic effects showed that all 5 PGRs induced malformations of the second antenna, rostrum, Malpighian tube, sensory bristles, and tail spine as well as function loss and death.
Related Materials