EMAIL THIS PAGE TO A FRIEND
Annales de dermatologie et de venereologie
[Toxidermatosis caused by sulfaguanidine].
PMID 2151594
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32402
Sulfaguanidine, VETRANAL®, analytical standard
C7H10N4O2S
S8751
Sulfaguanidine
C7H10N4O2S