EMAIL THIS PAGE TO A FRIEND
Synthesis
Suzuki-Miyaura cross-coupling of 3-pyridyl triflates with 1-alkenyl-2-pinacol boronates.
PMID 21516181
Abstract
Palladium-catalyzed Suzuki-type couplings of 3-pyridyl triflates with alkenyl pinacol boronates proceed in good to excellent yield. Optimized conditions use Pd(PPh(3))(4) (10 mol %) as catalyst with K(3)PO(4) (3 equiv) as base in dioxane.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
144428
2,6-Lutidine-α2,3-diol, 99%
C7H9NO2