EMAIL THIS PAGE TO A FRIEND
Asian Pacific journal of cancer prevention : APJCP
Genotoxic monitoring and benzene exposure assessment of gasoline station workers in metropolitan Bangkok: sister chromatid exchange (SCE) and urinary trans, trans-muconic acid (t,t-MA).
PMID 21517262
Abstract
Early warning of the potential of mutagens or carcinogens caused by benzene exposure that might occur in gasoline station workers can be achieved by examining 2 major biomarkers: sister chromatid exchange (SCE) and trans, trans-muconic acid (t,t-MA), a urinary metabolite of benzene. The main objective of this study was to assess benzene exposure and monitor the genotoxic effect of gasoline station workers in Bangkok, Thailand. Blood and urine samples were collected from 33 gasoline station workers, working in Pathumwan district area, central Bangkok, Thailand, for SCE and t,t-MA analysis, from April to June 2009. Control samples were collected from 30 office workers and students in the same area at the same period. Our results indicated significantly higher frequencies of SCE in gasoline exposed workers were than in controls (p<0.01), independent of gender. Urinary t,t-MA and t,t-MA/creatinine levels of gasoline exposed workers were also significantly higher than the control groups (p<0.05) were significantly higher in women than men workers (p<0.01). Calculated chromosomal damage relative risk (RR) of gasoline station workers was 3.00 (95% CI = 1.81 - 4.98, p<0.001) compared to controls. The gasoline exposed workers had potentially higher risk of chromosomal damage and cancer development because of direct contact to benzene.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M90003
trans,trans-Muconic acid, 98%
C6H6O4