EMAIL THIS PAGE TO A FRIEND
Gastroenterology
Which way to die: the regulation of acinar cell death in pancreatitis by mitochondria, calcium, and reactive oxygen species.
PMID 21524653
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T7515
Sodium taurolithocholate
C26H44NNaO5S