EMAIL THIS PAGE TO A FRIEND
Medical science monitor : international medical journal of experimental and clinical research
Inhibition of peripheral blood neutrophil oxidative burst in periodontitis patients with a homeopathic medication Traumeel S.
PMID 21525811
Abstract
The anti-inflammatory effects of a homeopathic remedy, Traumeel S, have been observed in experimental and clinical studies; however, its antioxidant properties have not been elucidated. The aim of the present study was to evaluate the antioxidant effects of Traumeel S on peripheral blood neutrophils in patients with periodontitis. The study was performed using venous blood of 22 individuals with chronic periodontitis and 21 healthy subjects. The antioxidant effects of Traumeel S on the production of reactive oxygen species by unstimulated and stimulated with unopsonized E. coli neutrophils were investigated using luminol- and lucigenin-dependent chemiluminescence (CL). Polymorphonuclear leukocytes of periodontitis patients produced higher levels (p<0.01) of light output of lucigenin-dependent chemiluminescence and significantly reduced (p<0.01) light output of luminol-dependent chemiluminescence than analogous cells of healthy subjects. Highly diluted (10⁻⁴ of the stem solution) Traumeel S significantly (by approximately 50%) reduced superoxide-induced oxidation of lucigenin by unstimulated and stimulated with unopsonized E. coli polymorphonuclear leukocytes of periodontitis patients and had a tendency to intensify luminol-dependent chemiluminescence. Preincubation of the unstimulated and stimulated with unopsonized E. coli polymorphonuclear leukocytes of healthy subjects with Traumeel S exerts no inhibitory action on the luminol- and lucigenin-dependent chemiluminescence of the above-mentioned cells. This study indicates that Traumeel S may significantly reduce production of superoxide anion by unstimulated and stimulated peripheral blood polymorphonuclear neutrophils of periodontitis patients.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M8010
N,N′-Dimethyl-9,9′-biacridinium dinitrate, used as chemiluminescent reagent
C28H22N4O6