EMAIL THIS PAGE TO A FRIEND
Toxicology
Exposure to 1-bromopropane causes degeneration of noradrenergic axons in the rat brain.
PMID 21527306
Abstract
1-Bromopropane (1-BP) has been used as an alternative to ozone-depleting solvents. Previous studies showed that 1-BP is neurotoxic in animals and humans. In humans, exposure to 1-BP caused various neurological and neurobehavioral symptoms or signs including depressive or irritated mood. However, the neurobiological changes underlying the depressive symptoms induced by 1-BP remain to be determined. The depressive symptoms are thought to be associated with degeneration of axons containing noradrenaline and serotonin. Based on this hypothesis, the present study examined the effects of repeated exposure to 1-BP on serotonergic and noradrenergic axons. Exposure to 1-BP induced dose-dependent decreases in the density of noradrenergic axons in the rat prefrontal cortex, but no apparent change in the density of serotonergic axons. The results suggest that depressive symptoms in workers exposed to 1-BP are due, at least in part, to the degeneration of noradrenergic axons in the brain.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B78106
1-Bromopropane, 99%
C3H7Br