EMAIL THIS PAGE TO A FRIEND
Journal of controlled release : official journal of the Controlled Release Society
Supramolecular triblock copolymers controlled by the coiled-coil motif: a new tool for drug delivery.
PMID 21529582