EMAIL THIS PAGE TO A FRIEND
Journal of controlled release : official journal of the Controlled Release Society
Protein macromonomers for covalent immobilization and subsequent triggered release from hydrogels.
PMID 21529598
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
109606
Methacrylamide, 98%
C4H7NO