EMAIL THIS PAGE TO A FRIEND
Journal of controlled release : official journal of the Controlled Release Society
Mesoporous silicon microparticles as carriers for peptides.
PMID 21529616
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M8693 Melanotan II acetate salt, ≥95% (HPLC)
C50H69N15O9 · xC2H4O2