EMAIL THIS PAGE TO A FRIEND
Journal of controlled release : official journal of the Controlled Release Society
Photo-crosslinked poly(trimethylene carbonate)-fumarate/n-vinyl pyrrolidone networks for the controlled release of proteins.
PMID 21529642