EMAIL THIS PAGE TO A FRIEND
The Canadian veterinary journal = La revue veterinaire canadienne
Successful therapy of coumatetralyl rodenticide induced pericardial effusion with pericardiocentesis in a dog.
PMID 21532823
Abstract
A 5-year-old, intact male, golden retriever was presented with an acute onset of lethargy and respiratory distress. The dog was diagnosed as having rodenticide intoxication with pericardial effusion. Pericardiocentesis was successfully performed and was followed with a blood transfusion. This case suggests that rodenticide intoxication might cause pericardial effusion in dogs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45404
Coumatetralyl, PESTANAL®, analytical standard
C19H16O3