EMAIL THIS PAGE TO A FRIEND
Biodegradation
Removal of methyl parathion and tetrachlorvinphos by a bacterial consortium immobilized on tezontle-packed up-flow reactor.
PMID 21533773
Abstract
A tezontle-packed up-flow reactor (TPUFR) with an immobilized bacterial consortium for biological treatment of methyl-parathion and tetrachlorvinphos was evaluated. These organophosphate pesticides are widely used in Mexico for insect and mite control, respectively. With the aim of developing a tool for pesticide biodegradation, four flow rates (0.936, 1.41, 2.19, and 3.51xa0l/h) and four hydraulic residence times (0.313, 0.206, 0.133, and 0.083xa0h) were evaluated in a TPUFR. In the bioreactor, with an operating time of 8xa0h and a flow of 0.936xa0l/h, we obtained 75% efficiency in the removal of methyl-parathion and tetrachlorvinphos. Their adsorptions in the volcanic rock were 9% and 6%, respectively. It was demonstrated that the removal of pesticides was due to the biological activity of the immobilized bacterial consortium. We confirmed the decrease in toxicity in the treated effluent from the bioreactor through the application of acute toxicity tests on Eisenia foetida. Immobilization of a bacterial consortium using tezontle as a support is innovative and an economical tool for the treatment of mixtures of organophosphorus pesticide residues.