EMAIL THIS PAGE TO A FRIEND
Clinical chemistry
New saccharogenic determination of alpha-amylase in serum and urine.
PMID 215341
Abstract
I describe a new kinetic enzymatic saccharogenic method for assaying alpha-amylase in human serum and urine. alpha-Amylase liberates maltose from starch. This is successively acted on by alpha-glucosidase, mutarotase, and glucose dehydrogenase. The resulting conversion of NAD+ to NADH, measured at 340 nm, during a 20-min incubation reflects amylase activity. Endogenous glucose is destroyed before measurement of amylase activity is begun.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M0902 Maltose Epimerase from Lactobacillus sp.