EMAIL THIS PAGE TO A FRIEND
Behavioural Brain Research
Neonatal exposure to propofol affects BDNF but not CaMKII, GAP-43, synaptophysin and tau in the neonatal brain and causes an altered behavioural response to diazepam in the adult mouse brain.
PMID 21540061