EMAIL THIS PAGE TO A FRIEND
International journal of pharmaceutics
Physicochemical characterization of drug-loaded rigid and elastic vesicles.
PMID 21540095
Abstract
Ketorolac loaded rigid and elastic vesicles were prepared by sonication and the physicochemical properties of the drug loaded-vesicle formulations were examined. Rigid and elastic vesicles were prepared from the double chain surfactant sucrose-ester laurate (L-595) and the single chain surfactant octaoxyethylene-laurate ester (PEG-8-L). Sulfosuccinate (TR-70) was used as a negative charge inducer. Evaluation of the prepared vesicle was performed by dynamic light scattering, extrusion and by (1)H NMR (T(2) relaxation studies). The vesicles mean size varied between 90 and 150 nm. The elasticity of the vesicles was enhanced with increasing PEG-8-L/L-595 ratio, while an increase in loading of ketorolac resulted in a reduction in vesicle elasticity. (1)H NMR measurements showed that the molecular mobility of ketorolac was restricted, which indicates that ketorolac molecules were entrapped within the vesicle bilayers. The T(2) values of the aromatic protons of ketorolac increased gradually at higher PEG-8-L levels, indicating that ketorolac mobility increased in the vesicle bilayer. The chemical stability of ketorolac was dramatically improved in the vesicle formulation compared to a buffer solution. The strong interactions of ketorolac with the bilayers of the vesicles might be the explanation for this increased stability of ketorolac.
Related Materials