EMAIL THIS PAGE TO A FRIEND
Journal of biomedicine & biotechnology
Synthesis and evaluation of amino acid-based radiotracer 99mTc-N4-AMT for breast cancer imaging.
PMID 21541217
Abstract
This study was to develop an efficient synthesis of (99m)Tc-O-[3-(1,4,8,11-tetraazabicyclohexadecane)-propyl]-α-methyl tyrosine ((99m)Tc-N4-AMT) and evaluate its potential in cancer imaging. N4-AMT was synthesized by reacting N4-oxalate and 3-bromopropyl AMT (N-BOC, ethyl ester). In vitro cellular uptake kinetics of (99m)Tc-N4-AMT was assessed in rat mammary tumor cells. Tissue distribution of the radiotracer was determined in normal rats at 0.5-4 h, while planar imaging was performed in mammary tumor-bearing rats at 30-120 min. The total synthesis yield of N4-AMT was 14%. Cellular uptake of (99m)Tc-N4-AMT was significantly higher than that of (99m)Tc-N4. Planar imaging revealed that (99m)Tc-N4-AMT rendered greater tumor/muscle ratios than (99m)Tc-N4. N4-AMT could be synthesized with a considerably high yield. Our in vitro and in vivo data suggest that (99m)Tc-N4-AMT, a novel amino acid-based radiotracer, efficiently enters breast cancer cells, effectively distinguishes mammary tumors from normal tissues, and thus holds the promise for breast cancer imaging.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
120693
α-Methyl-DL-tyrosine, 98%
C10H13NO3
M8131
α-Methyl-L-tyrosine, ≥98% (TLC)
C10H13NO3