EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Enantioselective dichlorination of allylic alcohols.
PMID 21542622
Abstract
The development of an enantioselective allylic alcohol dichlorination catalyzed by dimeric cinchona alkaloid derivatives and employing aryl iododichlorides as chlorine sources is reported. Reaction optimization, exploration of the substrate scope, and a model for stereoinduction are presented.