EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Research on the adsorption property of supported ionic liquids for ferulic acid, caffeic acid and salicylic acid.
PMID 21543269
Abstract
In this paper, the preparation of new supported ionic liquids (SILs) composed of the N-methylimidazolium cation and the quinoline cation is described. They have been confirmed and evaluated by infrared spectroscopy, elemental analysis and thermogravimetric analysis. Six kinds of different SILs included SiO(2)·Im(+)·Cl(-), SiO(2)·Im(+)·BF(4)(-), SiO(2)·Im(+)·PF(6)(-), SiO(2)·Qu(+)·Cl(-), SiO(2)·Qu(+)·BF(4)(-) and SiO(2)·Qu(+)·PF(6)(-). The adsorption characteristics of ferulic acid (FA), caffeic acid (CA) and salicylic acid (SA) on SILs were investigated by static adsorption experiments. It was found that SiO(2)·Qu(+)·Cl(-) had excellent adsorption and desorption capacity to three tested phenolic compounds. The dynamic adsorption characteristics of FA, CA and SA on SiO(2)·Qu(+)·Cl(-) were also studied. The saturated adsorption capacity of FA, CA and SA using SiO(2)·Qu(+)·Cl(-) as adsorbent was 64.6 mg/g, 53.2 mg/g and 72.2 mg/g respectively. Using 70% ethanol as eluent, the saturated desorption efficiencies of FA, CA and SA were 97.2%, 90.3% and 96.5% respectively. Thus, SiO(2)·Qu(+)·Cl(-) had strong adsorption and separation capacity for FA, CA and SA.