EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Acylation of heteroaromatic amines: facile and efficient synthesis of a new class of 1,2,3-triazolo[4,5-b]pyridine and pyrazolo[4,3-b]pyridine derivatives.
PMID 21544037
Abstract
1,2,3-Triazolo[4,5-b]pyridines and pyrazolo[4,3-b]pyridines can be readily prepared via cyanoacetylation reactions of 5-amino-1,2,3-triazoles 1a,b and 4-amino- pyrazole 2 followed by subsequent cyclization of the formed cyanoacetamides. Reactions of amines 1a,b with a mixture of p-nitrophenylacetic acid and acetic anhydride under microwave irradiation conditions afforded the corresponding amides 15a,b that underwent cyclization to form 1,2,3-triazolo[4,5-b]pyridines 16a,b upon heating in DMF solutions containing sodium acetate. Reactions of 1a,b with active methylene compounds, including 17a-c, in the presence of zeolites as catalyst also afforded 1,2,3-triazolo[4,5-b]pyridine derivatives 20a-f via the intermediacy of triazole derivatives 19 and not 18.