EMAIL THIS PAGE TO A FRIEND
The Journal of dermatology
Case of Ramsay-Hunt syndrome associated with dysphagia.
PMID 21545507
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M9756
Methylcobalamin, vitamin B12 analog
C63H91CoN13O14P