EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Expanded redox accessibility via ligand substitution in an octahedral Fe6Br6 cluster.
PMID 21547292
Abstract
Oxidation of the nominally all-ferrous hexanuclear cluster ((H)L)(2)Fe(6) with six equivalents of ferrocenium in the presence of bromide ions results in a six-electron oxidation of the Fe(6) core to afford the nominally all-ferric cluster ((H)L)(2)Fe(6)Br(6). The hexabromide cluster is also structurally characterized in a 4+ core oxidation state. A structural comparison of these two clusters provides an insight into the Fe(6) core electronic structure.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
388297
Ferrocenium hexafluorophosphate, 97%
C10H10F6FeP
482358
Ferrocenium tetrafluoroborate, technical grade
C10H10BF4Fe