EMAIL THIS PAGE TO A FRIEND
Magnetic resonance in chemistry : MRC
Precision studies in supramolecular chemistry: a 1H NMR study of hydroxymethoxyacetophenone/β-cyclodextrin complexes.
PMID 21547943
Abstract
The association constants for the interactions of 2-hydroxy-4-methoxyacetophenone, 2-hydroxy-5-methoxyacetophenone, 2-hydroxy-6-methoxyacetophenone, 3-hydroxy-4-methoxyacetophenone and 4-hydroxy-3-methoxyacetophenone with β-cyclodextrin in water were measured by (1)H NMR and by isothermal titration calorimetry. Very good agreement was obtained between the different methods. The errors associated with the NMR method for measuring mM binding affinities were estimated to be 10-30%, and by isothermal titration calorimetry, 10-20%. Rotating frame nuclear Overhauser effect spectroscopy studies show that the solution phase host-guest complexes formed by β-cyclodextrin with these hydroxymethoxyacetophenone derivatives are not structurally well defined but that the hydroxymethoxyacetophenone derivatives are mostly associated with the narrow primary hydroxyl rim.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
679704
3′-Hydroxy-4′-methoxyacetophenone, 97%
C9H10O3