EMAIL THIS PAGE TO A FRIEND
Glia
Role of Müller cells in cone mosaic rearrangement in a rat model of retinitis pigmentosa.
PMID 21547953
Abstract
Retinitis pigmentosa (RP) is a type of inherited retinal degenerative disease, which leads to blindness. The primary pathological event of this disease is the death of rods because of genetic mutations. The S334ter-line-3 rat is a transgenic model developed to express a rhodopsin mutation similar to that found in RP. In this study, the rod's death triggered are organization of the cone mosaic into an orderly array of rings. Four observations were relevant to understand this reorganization. First, rods died in hot spots, which progressively increased as circular waves, leaving rod-less zones behind. Second, rings of cones formed around these zones. Third, remodeled Müller glia processes enveloped cones and filled the center of their rings. Zonula occludens-1 located between the photoreceptor inner segments and the apical processes of Müller cells showed in the rings. Fourth, these rings were formed before the onset of cone cell deaths and were maintained until late stages of RP. From these observations,we hypothesize that cone-Müller-cell interactions mediate and maintain the rings. A test of this hypothesis can be performed by injecting DL-a-aminoadipic acid (AAA), which is known to disrupt Müller cell metabolism. A single intravitreal injection of AAA at P50 disrupted the rings of cones 3 days after the injection. These findings indicate that the rearrangement of cones in rings is modulated by Müller cells in RP. Thus, if the relationship between photoreceptors and Müller glia is better understood, the latter could potentially be manipulated for effective neuroprotection or the restoration of normal cone arrays.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A0637
DL-2-Aminoadipic acid, ≥99%
C6H11NO4