EMAIL THIS PAGE TO A FRIEND
Expert opinion on therapeutic patents
Modulation of GDP-fucose level for generating proteins with reduced rate of fucosylation (WO2010141855).
PMID 21548846
Abstract
The application (WO2010141855) is in the field of glycobiology, and involves the control of the rate of fucosylation of proteins by exogenous factors. It aims at controlling the rate of protein fucosylation with inhibitors (drugs or nucleic acid antagonists) of enzymes involved in the synthesis of GDP-fucose. Mammalian cell lines were cultured in the presence of inhibitors, for example, siRNA. The rates of GDP-fucose in cells and during protein fucosylation were characterized. The level of protein fucosylation decreases rapidly in response to a decrease in GDP-fucose level. The relationship between the rate of fucosylation of proteins and the level of GDP-fucose in a cell is non-linear. Reduction in the rate of protein fucosylation can be achieved with a minimal reduction of the level of GDP-fucose in cells. The paradigm may be used to synthesize proteins and antibodies, with a reduced rate of fucosylation. The application claims that the use of drugs or nucleic acid antagonists that inhibit the enzymes involved in GDP-fucose biosynthesis optimizes the level of GDP-fucose present in cells, and reduces the rate of fucosylation of glycoproteins.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G4401
Guanosine 5′-diphospho-β-L-fucose sodium salt, ≥85%
C16H23N5Na2O15P2