EMAIL THIS PAGE TO A FRIEND
Journal of bacteriology
Pantethine rescues phosphopantothenoylcysteine synthetase and phosphopantothenoylcysteine decarboxylase deficiency in Escherichia coli but not in Pseudomonas aeruginosa.
PMID 21551303
Abstract
Coenzyme A (CoA) plays a central and essential role in all living organisms. The pathway leading to CoA biosynthesis has been considered an attractive target for developing new antimicrobial agents with novel mechanisms of action. By using an arabinose-regulated expression system, the essentiality of coaBC, a single gene encoding a bifunctional protein catalyzing two consecutive steps in the CoA pathway converting 4'-phosphopantothenate to 4'-phosphopantetheine, was confirmed in Escherichia coli. Utilizing this regulated coaBC strain, it was further demonstrated that E. coli can effectively metabolize pantethine to bypass the requirement for coaBC. Interestingly, pantethine cannot be used by Pseudomonas aeruginosa to obviate coaBC. Through reciprocal complementation studies in combination with biochemical characterization, it was demonstrated that the differential characteristics of pantethine utilization in these two microorganisms are due to the different substrate specificities associated with endogenous pantothenate kinase, the first enzyme in the CoA biosynthetic pathway encoded by coaA in E. coli and coaX in P. aeruginosa.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P2125
D-Pantethine, analytical standard
C22H42N4O8S2
P3280
D-Pantethine hydrate, syrup
C22H42N4O8S2 · xH2O