EMAIL THIS PAGE TO A FRIEND
Journal of pharmacological sciences
Myosin light chain kinase/actin interaction in phorbol dibutyrate-stimulated smooth muscle cells.
PMID 21551973
Abstract
Previous work has suggested that in addition to its kinase activity, myosin light chain kinase (MLCK) exhibits non-kinase properties within its N-terminus that could influence cytoskeletal organization of smooth muscle cells (A. Nakamura et al. Biochem Biophys Res Commun. 2008;369:135-143). Myosin ATPase activity measurements indicate that the 26-41 peptide of MLCK significantly decreases ATPase activity as the concentration of this peptide increases. Sliding velocity of actin-filaments on myosin and stress responses in skinned smooth muscle tissue are also inhibited. Peptide-mediated uptake and the microinjection technique in cells indicate that the peptide was necessary for actin-filament stabilization. Fluorescence resonance energy transfer analysis indicated that in the presence of MLCK, α-actin but not β-actin remodeled during phorbol 12,13-dibutyrate (PDBu)-induced contractions. PDBu also induced podosomes in the cell. When MLCK expression was down-regulated by introduction of RNAi for MLCK by lentivirus vector into the cells, we failed to observe the podosome induction upon PDBu stimulation. Rescue experiments indicate that the non-kinase activity of MLCK plays an important role in maintaining actin stress fibers and in the PDBu-induced reorganization of actin-filaments in smooth muscle cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P1269
Phorbol 12,13-dibutyrate, ≥98% (TLC), powder
C28H40O8