Description
Molecular Formula
Add to Cart
07-355
Anti-acetyl-Histone H3 (Lys23) Antibody, Use Anti-acetyl-Histone H3 (Lys23) Antibody (Rabbit Polyclonal Antibody) validated in ChIP, WB to detect acetyl-Histone H3 (Lys23) also known as H3K23Ac, Histone H3 (acetyl K23).
07-360
Anti-acetyl-Histone H3 (Lys27) Antibody, Anti-acetyl-Histone H3 (Lys27) Antibody is a rabbit polyclonal antibody for detection of Histone H3 acetylated on lysine 27. Also known as Anti-H3K27ac this antibody is published in peer reviewed journals and is specificity verified by dot blot (DB) and validated in ChIP, ChIP-seq, WB, DB, Mplex.
06-866
Anti-acetyl-Histone H4 Antibody, Use Anti-acetyl-Histone H4 Antibody (Rabbit Polyclonal Antibody) has been published and validated in ChIP, WB, ICC to detect acetyl-Histone H4 also known as H4 histone family member A, histone 1-H4a.
07-595
Anti-acetyl-Histone H4 (Lys12) Antibody, Use Anti-acetyl-Histone H4 (Lys12) Antibody (Rabbit Polyclonal Antibody) validated in ChIP, DB, WB, ChIP-seq to detect acetyl-Histone H4 (Lys12) also known as H4K12Ac, Histone H4 (acetyl K12).
07-327
Anti-acetyl-Histone H4 (Lys5) Antibody, Use Anti-acetyl-Histone H4 (Lys5) Antibody (Rabbit Polyclonal Antibody) validated in WB, DB, ICC, ChIP-seq, ChIP to detect acetyl-Histone H4 (Lys5) also known as H4K5Ac, Histone H4 (acetyl K5).
07-328
Anti-acetyl-Histone H4 (Lys8) Antibody, Use Anti-acetyl-Histone H4 (Lys8) Antibody (Rabbit Polyclonal Antibody) validated in ChIP, IP, WB, DB, ChIP-seq to detect acetyl-Histone H4 (Lys8) also known as H4K8Ac, Histone H4 (acetyl K8).
07-274
Anti-dimethyl-Histone H3 (Lys36) Antibody, Use Anti-dimethyl-Histone H3 (Lys36) Antibody (Rabbit Polyclonal Antibody) validated in WB, DB to detect dimethyl-Histone H3 (Lys36) also known as H3K36me2, Histone H3 (di methyl K36).
07-030
Anti-dimethyl-Histone H3 (Lys4) Antibody, Use Anti-dimethyl-Histone H3 (Lys4) Antibody (rabbit polyclonal antibody) is published and validated in ChIP, DB, ICC, IF, WB, PIA, ChIP-seq. to detect dimethyl-Histone H3 (Lys4) also known as H3K4me2, Histone H3 (di methyl K4).
07-441
Anti-dimethyl-Histone H3 (Lys9) Antibody, Use Anti-dimethyl-Histone H3 (Lys9) Antibody (Rabbit Polyclonal Antibody) validated in DB, ICC, PIA, WB to detect dimethyl-Histone H3 (Lys9) also known as H3K9me2, Histone H3 (di methyl K9).
07-448
Anti-monomethyl-Histone H3 (Lys27) Antibody, Anti-monomethyl-Histone H3 (Lys27) Antibody is a rabbit polyclonal antibody to detect Histone H3 monomethylated at lysine 27. This purified Ab, also known as Anti-H3K27me1 is specificity verified by dot blot (DB), published in peer reviewed journals and validated in ICC, WB, ChIP, ChIP-seq.
07-449
Anti-trimethyl-Histone H3 (Lys27) Antibody, Anti-trimethyl-Histone H3 (Lys27), also known as Anti-H3K27me3, is a highly published Rabbit Polyclonal Antibody. This protein A purified antibody is dot blot tested for trimethylated lysine 27 specificity and validated in WB, ChIP, ChIP-seq, ICC, IP.
07-473
Anti-trimethyl-Histone H3 (Lys4) Antibody, Anti-trimethyl-Histone H3 (Lys4) Antibody is a rabbit polyclonal antibody for detection of Histone H3 trimethylated at lysine 4. Also known as Anti-H3K4me3, this highly specific and well published antibody has been validated in ChIP, DB, WB, PIA, ChIP-seq.